EP0030123301
Eppendorf® Safe-Lock micro test tubes
capacity 0.5 mL, PCR Clean, colorless, pack of 500 ea
Select a Size
430,00 kr.
Select a Size
About This Item
430,00 kr.
Recommended Products
material
cap (plug seal)
colorless
colorless cap
colorless tube
conical bottom
sterility
non-sterile
feature
graduations
neck thread Push fit
packaging
pack of 500 ea
manufacturer/tradename
Eppendorf® 0030123301
capacity
0.5 mL
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- The hinged Eppendorf® Safe-Lock lid provides additional protection against unintentional lid opening during incubation and storage.
- g-Safe®:Provides great centrifugation stability and prevents sample loss due to tube breakage
- The optimized sealing properties ensures safety when working with hazardous samples and minimal evaporation rates during long-term storage.
- The large frosted lid and wide side-surface area make labeling convenient.
- Functions between temperatures -86 °C to 100 °C
- Autoclavable when open (121 °C, 20 min)
- Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage.
- Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes.
- Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results, g-Safe®: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples Precise lid sealing for minimal evaporation rates during long-term storage.
- Large frosted lid and surface on the side for easy labelling.
- Guaranteed functionality from -86 °C to 100 °C.
- Autoclavable when open (121 °C, 20 min).
- Available in Eppendorf Protein and DNA LoBind material for maximum recovery of valuable samples.
Using the Eppendorf 30,000 × g system comprising of Safe-Lock Tubes and Centrifuge 5430 R with high-speed rotor, the effect of relative centrifugal force (rcf), duration of centrifugation and the amount of isopropanol on the recovery rate of plasmid DNA from alcohol precipitation were investigated. All three factors play a role, with rcf having the greatest effect. By centrifuging the Eppendorf Safe-Lock Tubes at 30,000 × g, nearly 90% of DNA could be recovered from a 5 min centrifugation.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service