EMU209151
MISSION® esiRNA
targeting mouse Ensmusg00000060559
Select a Size
1.360,00 kr.
Select a Size
About This Item
1.360,00 kr.
description
Powered by Eupheria Biotech
Quality Level
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
CAGAGATTATTATGCCTTCTGAAAAACTTTAACCTTGTTTCTGTGCGAAGTTCTGATTTGCATAGAAGCTCTAAAGAGACATAAGAAGCTCAGGGTGTAGTTGAGTTTGCCTCAAGATCACCAGGAACACTTGATTGTGGTCATGAGATGGCTCAGCAGATAAAGGCGTTTGCTGCCAAGCCTGGGTATTTGAGTATGATCCCATGAACCCACGTGGTGAGAGAAGAGGACTGACTTTGAGAGGCGCGCGCGCGCGCACACACACACACACACACACACACACAGAGAGAGAGAGAGAGAGAGAGAGAGAGAGAGAGAGAGAGAGAGAAGTGTTTTTTAATAGATAATTGGGCACCACTCCTAGGTTCTATCTCAAATGATTGAGTTAGTATAGAGAGAGGCATCTAACTCGTTTGTCATTAGAAAGAAAATAATCCCATGTACTTATTTTGTGTGAATGTGCAGGAAAGTGGAAGTCAGGAGATATCAGAAGACAGTTTATGCTCTCCA
shipped in
ambient
storage temp.
−20°C
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service