Skip to Content
Merck
All Photos(1)

Key Documents

EHU144411

Sigma-Aldrich

MISSION® esiRNA

targeting human ARF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTTAAGCTGGGTGAGATCGTGACCACCATTCCCACCATAGGCTTCAACGTGGAAACCGTGGAGTACAAGAACATCAGCTTCACTGTGTGGGACGTGGGTGGCCAGGACAAGATCCGGCCCCTGTGGCGCCACTACTTCCAGAACACACAAGGCCTGATCTTCGTGGTGGACAGCAATGACAGAGAGCGTGTGAACGAGGCCCGTGAGGAGCTCATGAGGATGCTGGCCGAGGACGAGCTCCGGGATGCTGTCCTCCTGGTGTTCGCCAACAAGCAGGACCTCCCCAACGCCATGAATGCGGCCGAGATCACAGACAAGCTGGGGCTGCACTCACTACGCCACAGGAACTGGTACATTCAGGCCACCTGCGCCACCAGCGGCGACGGGCTCTATGAAGGACTGGACTGGCTGTCCAATCAGCTCCGGAACCAGAAGTGAACGCGACCCCCCTCCCTCTCACTCCTCTTGCCCTCTGCTTTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nisha Bte Mohd Rafiq et al.
The Journal of cell biology, 216(1), 181-197 (2016-12-23)
Podosomes represent a class of integrin-mediated cell-matrix adhesions formed by migrating and matrix-degrading cells. We demonstrate that in macrophage-like THP1 cells and fibroblasts stimulated to produce podosomes, down-regulation of the G-protein ARF1 or the ARF1 guanine nucleotide exchange factor, ARNO
Tatsuyuki Matsudaira et al.
Journal of cell science, 128(16), 3131-3142 (2015-07-03)
The retrograde pathway is defined by the transport of proteins and lipids from the plasma membrane through endosomes to the Golgi complex, and is essential for a variety of cellular activities. Recycling endosomes are important sorting stations for some retrograde
Christin Münzberg et al.
Journal of cellular and molecular medicine, 19(5), 948-959 (2015-03-11)
Hypersecretion is the major symptom of functional neuroendocrine tumours. The mechanisms that contribute to this excessive secretion of hormones are still elusive. A key event in secretion is the exit of secretory products from the Golgi apparatus. ADP-ribosylation factor (Arf)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service