Skip to Content
Merck
All Photos(1)

Key Documents

EHU111901

Sigma-Aldrich

MISSION® esiRNA

targeting human NLRX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCGGTGTCAGAATACTGGTCAGTGATCCTCAGTGAAGTCCAGCGGAACCTCAATAGCTGGGATCGGGCCCGGGTTCAGCGACACCTTGAGCTCCTACTGCGGGATCTGGAAGATAGCCGGGGTGCCACCCTTAATCCTTGGCGCAAGGCCCAGCTGCTGCGAGTGGAGGGCGAGGTCAGGGCCCTCCTGGAGCAGCTGGGAAGCTCTGGAAGCTGAGACACTGGCGGCAGGCACCTAGCTATGTGACCACTGGCCCTAAACCTTTTCCCTCTGTGGCCTCCTGGCTTGCACTGCTCCCTCTAGAAAGATTCCTTCAGGTCTGGAGGCAGAGGAATGGGCATAGCTGAGCCAGTTGCCCTCCTAGGGCATGTTTGACCAGGACTGAGTCTGGAATCTCCAAGTTAAAGATGGTGAATCAATGCTTCGGGCTTGGAGATGGAACATGCCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ding Ma et al.
International immunopharmacology, 71, 7-13 (2019-03-13)
Osteoarthritis (OA) is a chronic debilitating disease characterized by joint degeneration. Excessive chondrocyte apoptosis and inflammation contributes to articular cartilage destruction in OA pathology. Nucleotide-binding oligomerization domain (NOD)-like receptor X1 (NLRX1) has emerged as a critical regulator of inflammation that
Haiyan Yin et al.
Scientific reports, 7, 44311-44311 (2017-03-14)
Nucleotide-binding domain and leucine-rich-repeat-containing family member X1 (NLRX1), located in mitochondria, can recognize cytoplasmic pattern recognition receptors and is tightly related to reactive oxygen species (ROS) production, mitochondrial function, apoptosis and inflammation. The present study was designed to explore whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service