Skip to Content
Merck
All Photos(1)

Key Documents

EHU078901

Sigma-Aldrich

MISSION® esiRNA

targeting human GPI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CACCAAGGCACCAAGATGATACCCTGTGACTTCCTCATCCCGGTCCAGACCCAGCACCCCATACGGAAGGGTCTGCATCACAAGATCCTCCTGGCCAACTTCTTGGCCCAGACAGAGGCCCTGATGAGGGGAAAATCGACGGAGGAGGCCCGAAAGGAGCTCCAGGCTGCGGGCAAGAGTCCAGAGGACCTTGAGAGGCTGCTGCCACATAAGGTCTTTGAAGGAAATCGCCCAACCAACTCTATTGTGTTCACCAAGCTCACACCATTCATGCTTGGAGCCTTGGTCGCCATGTATGAGCACAAGATCTTCGTTCAGGGCATCATCTGGGACATCAACAGCTTTGACCAGTGGGGAGTGGAGCTGGGAAAGCAGCTGGCTAAGAAAATAGAGCCTGAGCTTGATGGCAGTGCTCAAGTGACCTCTCACGACGCTTCTACC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Zong et al.
Arthritis research & therapy, 17, 100-100 (2015-04-19)
Fibroblast-like synoviocytes (FLS) play an important role in the pathogenesis of rheumatoid arthritis (RA). This study aimed to investigate the role of glucose 6-phosphate isomerase (GPI) in the proliferation of RA-FLS. The distribution of GPI in synovial tissues from RA
Yiran Li et al.
International journal of oncology, 47(3), 1017-1024 (2015-07-24)
Autocrine motility factor (AMF) as a cytokine and a growth factor, is known to regulate tumor cell growth and motility in the progress of various human malignant tumors, however, its role in endometrial cancer (EC) has not been fully studied.
Dongling Zou et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(9), 6725-6732 (2015-04-03)
Chemotherapy is the preferred therapeutic approach for the therapy of advanced ovarian cancer, but 5-year survival rate remains low due to the development of drug resistance. Increasing evidence has documented that microRNAs (miRNAs) act important roles in drug resistance in
Gina Shaw-Hallgren et al.
PloS one, 9(5), e96506-e96506 (2014-05-13)
Nemo-like kinase (NLK), a proline-directed serine/threonine kinase regulated by phosphorylation, can be localized in the cytosol or in the nucleus. Whether the localization of NLK can affect cell survival or cell apoptosis is yet to be disclosed. In the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service