Skip to Content
Merck
All Photos(1)

Key Documents

EHU031761

Sigma-Aldrich

MISSION® esiRNA

targeting human TRPC3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGGGTCTGCTTGTGTTCAATGCCTCAGACAGGTTCGAAGGCATCACCACGCTGCCCAATATCACAGTTACTGACTATCCCAAACAGATCTTCAGGGTGAAAACCACCCAGTTTACATGGACTGAAATGCTAATTATGGTCTGGGTTCTTGGAATGATGTGGTCTGAATGTAAAGAGCTCTGGCTGGAAGGACCTAGGGAATACATTTTGCAGTTGTGGAATGTGCTTGACTTTGGGATGCTGTCCATCTTCATTGCTGCTTTCACAGCCAGATTCCTAGCTTTCCTTCAGGCAACGAAGGCACAACAGTATGTGGACAGTTACGTCCAAGAGAGTGACCTCAGTGAAGTGACACTCCCACCAGAGATACAGTATTTCACTTATGCTAGAGATAAATGGCTCCCTTCTGACCCTCAGATTATATCTGAAGGCCTTTATGCCATAGCTGTTGTGCTCAGCTTCTCTCGGATTGCGTAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingwei Wang et al.
Biochemical and biophysical research communications, 484(1), 209-217 (2016-12-31)
Airway hyperresponsiveness (AHR), airway remodeling and inflammation are the fundamental pathological alterations that occur in asthma. Transient receptor potential canonical 3 (TRPC3) has been implicated in diverse functions of airway smooth muscle cells (ASMCs) in asthma. However, the underlying mechanisms
Xiao-Xu Chen et al.
Life sciences, 187, 64-73 (2017-08-15)
Canonical transient receptor potential channel-3 (TRPC3)-encoded Ca Primary mouse ASMCs were cultured with or without ACh treatment, then cell viability, TRPC3 expression, NSCC currents and [Ca TRPC3 blocker Gd Our data suggested ACh could induce ASMC proliferation, and TRPC3 may
Pengzhou Hang et al.
International journal of biological sciences, 11(5), 536-545 (2015-04-22)
Brain-derived neurotrophic factor (BDNF) is associated with coronary artery diseases. However, its role and mechanism in myocardial infarction (MI) is not fully understood. Wistar rat and Kunming mouse model of MI were induced by the ligation of left coronary artery.
Xiaoyu Zhang et al.
Journal of cellular biochemistry, 119(7), 6033-6044 (2018-03-27)
This study aimed to validate whether transient receptor potential channel1 (TRPC1) and TRPC3 participate in the regulation the proliferation of airway smooth muscle cells (ASMCs) through modulating calcium ion (Ca2+ ) influx in vitro. Chronic model of murine asthma was
Kexin Meng et al.
PloS one, 9(6), e98777-e98777 (2014-06-07)
Calcium-sensing receptor (CaSR) has been demonstrated to be present in several tissues and cells unrelated to systemic calcium homeostasis, where it regulates a series of diverse cellular functions. A previous study indicated that CaSR is expressed in mouse glomerular mesangial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service