Skip to Content
Merck
All Photos(1)

Key Documents

D9288

Sigma-Aldrich

1,6-Dinitropyrene

~60% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N2O4
CAS Number:
Molecular Weight:
292.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

concentration

~60% (TLC)

mp

>300 °C (lit.)

storage temp.

−20°C

SMILES string

[O-][N+](=O)c1ccc2ccc3c(ccc4ccc1c2c34)[N+]([O-])=O

InChI

1S/C16H8N2O4/c19-17(20)13-8-4-10-2-6-12-14(18(21)22)7-3-9-1-5-11(13)16(10)15(9)12/h1-8H

InChI key

GUXACCKTQWVTLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B A Smith et al.
Mutation research, 379(1), 61-68 (1997-10-23)
Direct pulmonary instillation of 1,6-dinitropyrene (DNP) into male Fischer 344 rats results in a dose-dependent induction of lung tumors and 6-thioguanine-resistant (TGr) T-lymphocytes. The treatment also results in DNP binding to dG in the lung and in T-lymphocytes. In the
Nitroarenes (selected) 1,6-dinitropyrene.
Report on carcinogens : carcinogen profiles, 11, III186-III187 (2004-01-01)
Akiko H Hashimoto et al.
Environmental and molecular mutagenesis, 47(4), 277-283 (2006-02-21)
1,6-Dinitropyrene (1,6-DNP) is a ubiquitous airborne pollutant found in diesel exhaust. In this study, mutagenesis was examined in the lungs of gpt-delta transgenic mice after intratracheal instillation of 0-0.1 mg 1,6-DNP. In addition, the 1,6-DNP-induced gpt mutation spectrum was compared
F Kiefer et al.
Toxicology letters, 96-97, 131-136 (1998-11-20)
The effect of caffeine on the genotoxicity of several carcinogenic compounds representing various classes of chemicals was investigated in a panel of V79 Chinese hamster cells genetically engineered to express cytochromes P4501A1 and P4501A2 and differing in their expression of
I Tahara et al.
Mutation research, 343(2-3), 109-119 (1995-06-01)
This study examined the stability of mutagenic 1-nitropyrene (1-NP) and (1,6-dinitropyrene (1,6-diNP) in environmental water samples and various soil suspensions containing 0.1% peptone and in water samples containing no peptone. The water samples or the soil suspensions were mixed with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service