D3750
Dehydrocholic acid
Synonym(s):
(5β)-3,7,12-Trioxocholan-24-oic acid, 3,7,12-Trioxo-5β-cholanic acid, 5β-Cholanic acid-3,5,12-trione
About This Item
Recommended Products
description
anionic
mp
238-240 °C
solubility
ethanol: 10 mg/mL
SMILES string
[H][C@@]12CC(=O)CC[C@]1(C)[C@@]3([H])CC(=O)[C@]4(C)[C@H](CC[C@@]4([H])[C@]3([H])C(=O)C2)[C@H](C)CCC(O)=O
InChI
1S/C24H34O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-14,16-18,22H,4-12H2,1-3H3,(H,28,29)/t13-,14+,16-,17+,18+,22+,23+,24-/m1/s1
InChI key
OHXPGWPVLFPUSM-KLRNGDHRSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
replaced by
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service