Skip to Content
Merck
All Photos(5)

Key Documents

CMC0005

Sigma-Aldrich

SIG10 F′ MAX Electrocompetent Cells

for general cloning & library production

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

grade

for molecular biology

form

buffered aqueous solution

cell transformation

competent cell type: electrocompetent
transformation efficiency: ≥2 x 1010 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

The SIG10 F′ MAX Electrocompetent Cells provide large numbers of transformants from hard-to-clone fragments or limited DNA at a lower price.

Genotype

[F´ pro A+B+ lacIqZΔM15::Tn10 (TetR)] /mcrA Δ(mrr-hsdRMS-mcrBC) endA1 recA1 Φ80dlacZΔM15 ΔlacX74 araD139 Δ(ara, leu)7697 galU galK rpsL nupG λ- tonA (StrR)

Features and Benefits

  • share the most useful genetic elements of standard cloning strains like DH5α DH10B, JM109, TOP10, etc. and directly replace them in cloning protocols.
  • provide the performance researchers need with ease of use.
  • provide solutions for a wide range of applications at economical prices.
  • Blue - white screening

Components

  • SIG10 F′ MAX electrocompetent cells
  • pUC 19 transformation control DNA
  • recovery medium for cloning


Legal Information

DH5α is a trademark of Invitrogen Corp.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service