Skip to Content
Merck
All Photos(1)

Key Documents

CLS4840

Corning® microvolume pipet tips

0.2-10 μL, universal fit bulk packed pipet tips, natural, non-sterile, 10,000 tips/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

10000 EA
4.320,00 kr.

4.320,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
10000 EA
4.320,00 kr.

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
0.2—10 μL
sterility:
non-sterile
feature:
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation
packaging:
bag of 1000
case of 10000
manufacturer/tradename:
Corning 4840

4.320,00 kr.


Please contact Customer Service for Availability

material

natural tip
polypropylene

sterility

non-sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation

packaging

bag of 1000
case of 10000

manufacturer/tradename

Corning 4840

volume range

0.2-10 μL

volume

0.2—10 μL

compatibility

for use with Gilson® and other popular ultra-micropipettors

Looking for similar products? Visit Product Comparison Guide

General description

Microvolume Tips provide accurate, reliable performanace for major brand ultra-pipettors. All racked tips are certified RNase/DNase-free and nonpyrogenic.

Legal Information

Corning is a registered trademark of Corning, Inc.
Gilson is a registered trademark of Gilson, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service