CLS480134
Corning® Microcentrifuge Individual Adapters
Synonym(s):
lab equipment accessory
Select a Size
423,00 kr.
Select a Size
About This Item
423,00 kr.
feature
Holds 0.5/0.4 mL tubes
packaging
pack of 6 ea
manufacturer/tradename
Corning 480134
technique(s)
centrifugation: suitable
compatibility
for use with 6765-HS
for use with 6766-HS
for use with 6767-HS
for use with Corning® LSE High Speed Microcentrifuges
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service