Key Documents
CLS4519156
Corning® ProCulture® spinner flask accessories
side arm fitting, Dual with 1/4 inlets for 6 L spinner flask, Gas and Media Handling Fitting with Dual 1/4 in. Inlets for 6L Angled Sidearm Flasks
Synonym(s):
container accessory
Select a Size
1 EA
4.730,00 kr.
4.730,00 kr.
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
4.730,00 kr.
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
4.730,00 kr.
Please contact Customer Service for Availability
manufacturer/tradename
Corning 4519-156
General description
Side arm fittings used for a variety of functions in angled sidearm Spinner Flasks during cell culture. Sidearm fittings provide a convenient way of transferring media into or out of flasks aseptically. They can also be used to sparge the cell culture medium with gases to enhance cell production. Fittings comprise of a PET insert with a Viton O-ring and a polypropylene sealing cap & 316 stainless steel tubes.
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Legal Information
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)