CLS431079
Corning® cell culture flasks
surface area 175 cm2, angled neck, cap (plug seal)
Synonym(s):
culture flask, flask cell culture, tissue culture flask
About This Item
Recommended Products
material
U-bottom flask
angled neck
cap (plug seal)
plug-seal screw-cap
polystyrene
rectangular flask
sterility
sterile
feature
cap
graduations
packaging
case of 50
manufacturer/tradename
Corning 431079
maximum volume
250 mL
surface area
175 cm2
working volume
35-52.5 mL
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- Manufactured from optically clear virgin polystyrene
- Treated for optimal cell attachment
- Printed with lot numbers for ease in traceability
- 100% integrity tested
- Sterilized by gamma irradiation and certified nonpyrogenic
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service