CLS354539
Corning® BioCoat™ Poly-D-Lysine Cell Culture Flask
capacity 175 cm2, rectangular flask, Straight neck, vented, Cap
Synonym(s):
cell culture flask, plastic cell culture flask, coated flask
Select a Size
1.160,00 kr.
Select a Size
About This Item
1.160,00 kr.
Recommended Products
material
rectangular flask
feature
vented Cap
Straight neck
packaging
pkg of 5 ea (1 pack of 5)
manufacturer/tradename
Corning 354539
capacity
175 cm2
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service