CLS21505L
Pyrex® single cavity chromatography reservoir
capacity 5 L
Synonym(s):
storage reservoir
About This Item
Recommended Products
material
Pyrex
colorless
grade
for chromatography
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Corning 2150-5L
capacity
5 L
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service