Skip to Content
Merck
All Photos(1)

Documents

MH-22A

96121721, mouse liver, Polymorphic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MH-22A, 96121721

biological source

mouse liver

growth mode

Adherent

karyotype

2n = 40

morphology

Polymorphic

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C3HA hepatocarcinoma

Cell Line Description

MH-22A was derived from the cells of a solid form of mouse hepatoma 22A. Inoculation into C3HA mice led to the formation of transplantable tumours.

Application

Experimental tumour therapy in vitro and in vivo

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:5 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service