Skip to Content
Merck
All Photos(1)

Key Documents

TFT B1

94102701, Reactivity: Anti-Ricin β-chains

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/5 (clone of SP2/0 Ag-14)

Reactivity

Anti-Ricin β-chains

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/5 x Mouse BALB/c spleen immunised with ricin ß chain

Cell Line Description

Produces a monoclonal antibody (IgG1) which reacts with the ß chain of the plant toxin ricin.

Immunogen

Ricin ß chain

Application

Has been used in a protocol for purifying the α chains (A1 and A2) of ricin with minimal cross-contamination for the preparation of immunotoxins.

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 100,000 - 1,000,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service