KB (HeLa derivative)
94050408, human cervix, Epithelial-like
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
KB (HeLa derivative), 94050408
biological source
human cervix
growth mode
Adherent
karyotype
2n = 46
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human cervix carcinoma
Cell Line Description
Originally derived from an epidermoid carcinoma of the mouth of an adult male Caucasian. It was one of the early attempts to isolate and serially propagate a human cell line directly on glass as a monolayer. NB: This cell line contains HeLa marker chromosomes and expresses type A G6PD. For further information see Nature 1976;259:211; In Vitro 1978;14:469. This cell line was found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cell line should be considered as derived from HeLa. Ethnicity: Black.
Application
KB has been used extensively in studies of cell nutrition and metabolism, cancer chemotherapy screening, tumourigenicity and viruses e.g. susceptible to Polio virus type I and Adenovirus type 3.
Culture Medium
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service