Skip to Content
Merck
All Photos(1)

Documents

PORTHOS

93122323, Porcine thyroid, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

PORTHOS, 93122323

biological source

Porcine thyroid

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Thyroglobulin, plasminogen activator (u-PA)

receptors

Thyrotropin, insulin, Insulin Growth Factor (IGF-1), hydrocortisone

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Porcine thyroid epithelial

Cell Line Description

Derived from porcine thyroid. The cells have been adapted to grow in low serum (0.1%) media.

Application

Regulation of thyroid cell metabolism

Culture Medium

Click:RPMI (1:1) + 10μg/ml Insulin + 10mM Hydrocortisone + 5μg/ml Transferrin + 10ng/ml glycyl-L-histydyl-L-lysine acetate + 10ng/ml somatostatin + 10mU/ml Thyroid Stimulating Hormone (TSH) + 2mM Glutamine + 0.1% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 seeding at 2 x 10 4 cells/cm2 using 0.05% trypsin/0.025% EDTA in Puck F saline; 2.5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service