RENT RO3
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
rat endometrium
packaging
tube of 5 μg 92092514-DNA-5UG
pkg of vial of cells 92092514-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Spindle shaped
products
Not specified
receptors
Glucocorticoid
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat Sprague-Dawley, primary endometrial, SV40 T antigen transformed
Cell Line Description
Derived from the endometrium tissue from the uterus of a 21-28 day-old Sprague-Dawley rat. The cells carry vimentin, uvomorulin, proenkephalin, desmin and connexin 43 markers.
Culture Medium
DMEM + 20μg/l Gly-His-Lys (Sigma) + 2mM Glutamine + 80 IU human Insulin/l (Novo) + 10% Foetal Bovine Serum (FBS) (heat-inactivated).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:5 i.e. seeding at 2x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Doubling time 35 h.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service