Recommended Products
product name
G-402, 90112715
biological source
human kidney
growth mode
Adherent
karyotype
Modal no. 47
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Related Categories
Cell Line Origin
Human Caucasian renal leiomyoblastoma
Cell Line Description
Derived from a tumour of a 9 month old female Caucasian. Highly transformed and grows in soft agar.
Application
Transformation and tumourigenicity studies
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10,13
D13S317: 9,12
D16S539: 11,13
D5S818: 11,12
D7S820: 9,10
THO1: 7,9
TPOX: 10
vWA: 13,17
CSF1PO: 10,13
D13S317: 9,12
D16S539: 11,13
D5S818: 11,12
D7S820: 9,10
THO1: 7,9
TPOX: 10
vWA: 13,17
Culture Medium
McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Change medium every 4 days.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service