Skip to Content
Merck
All Photos(1)

Documents

P815-1-1

85011406, mouse breast, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

P815-1-1, 85011406

biological source

mouse breast

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse DBA/2 mastocytoma

Cell Line Description

From DBA/2 mouse tumour treated with methylcholanthrene. Used as target cell for cytotoxic T-cell assays. Cells exhibit no effector activity in antibody dependent cell mediated cytotoxic system. Growth not inhibited by dextran sulphate, LPS or PPD.

Application

Cytotoxicity and tumourigenicity studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml. The cells are non-adherent but can give rise to adherent cells with large granules; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service