Skip to Content
Merck
All Photos(1)

Key Documents

RB247C

6070601, human eye, Clusters of round cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

RB247C, 06070601

biological source

human eye

growth mode

Suspension

karyotype

Not specified

morphology

Clusters of round cells

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Retinoblastoma

Cell Line Description

Human retinoblastoma cell line derived from a male patient. Bilateral retinoblastoma, tumour treated by irradiation. Tumourigenicity not tested.

Culture Medium

Iscove′s Modified DMEM + 2mM Glutamine + 15% Foetal Bovine Serum (FBS) + 10ug/ml Insulin + 55uM 2-Mercaptoethanol

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 2-3 x100,000 cells/ml; 5% CO2; 37°C. Saturation density: 2 x 1000,000 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service