Skip to Content
Merck
All Photos(1)

Documents

Vero/hSLAM

4091501, African green monkey kidney, Fibroblast

Synonym(s):

Vero/hSLAM Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Vero/hSLAM, from African green monkey kidney, 04091501, Convention on International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

African green monkey kidney

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

SLAM; also known as CDw150, a receptor for measles virus

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Monkey African Green Kidney human CDw150 (SLAM) mealses virus receptor expression

Cell Line Description

Vero/hSLAM was derived by transfection of Vero cells with a vector plasmid (pCXN2) containing the neomycin resistance gene and an expression plasmid (pCAG-hSLAM) encoding the human signaling lymphocytic activation molecule (SLAM) , also known as CDw150, which is a receptor for measles virus. Clones of transfected cells were selected in the presence of geneticin (also known as G418). Vero/hSLAM cells can be used to isolate measles virus from human clinical samples and are a substitute for the B95a cell line for this purpose. Mumps susceptibility has also been demonstrated at the depositor′s laboratory.

Culture Medium

MEM with Earle's salts + 2 mM L-Glutamine (G7513) + 5% FBS / FCS (F2442) + 0.4 mg/ml Geneticin. It is essential that geneticin is added to the culture medium as it is required to maintain the expression of the SLAM gene.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:4, i.e., seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service