Skip to Content
Merck
All Photos(1)

Documents

C5223

Sigma-Aldrich

Anti-Cdk2 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous glycerol solution

Synonym(s):

Anti-CDKN2, Anti-p33(CDK2)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

rat (predicted), human, mouse (predicted)

technique(s)

immunoprecipitation (IP): 2-4 μg using 500 μg of HeLa cell nuclear extract
western blot: 0.5-2 μg/mL using human HeLa cell nuclear extract

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CDK2(1017)
mouse ... Cdk2(12566)

Specificity

The antibody reacts specifically with human cyclin-dependent kinase 2 (Cdk2) (p33cdk2).

Immunogen

synthetic peptide corresponding to amino acids 287-298 (CQDVTKPVPHLRL) of human cdk2.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Immunoprecipitation (1 paper)

Physical form

Solution in 0.1 M Tris-glycine, pH 7.4, 0.15 M sodium chloride, and 0.05% sodium azide containing 30% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonie W Wahjudi et al.
International journal of cancer, 148(6), 1438-1451 (2020-09-20)
DNA sequencing and RNA sequencing are increasingly applied in precision oncology, where molecular tumor boards evaluate the actionability of genetic events in individual tumors to guide targeted treatment. To work toward an additional level of patient characterization, we assessed the
Christoffer Bento et al.
BMC cell biology, 10, 89-89 (2009-12-19)
The DDIT3 gene encodes a transcription factor belonging to the CCAAT/enhancer binding protein (C/EBP) family. It is normally expressed at very low levels but is activated by cellular stress conditions and induces G1 arrest and, in some cell types, apoptosis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service