Skip to Content
Merck
All Photos(1)

Key Documents

C5040

Sigma-Aldrich

Clemizole hydrochloride

Synonym(s):

1-(p-Chlorobenzyl)-2-(1-pyrrolidinylmethyl)benzimidazole hydrochloride, 1-(p-Chlorobenzyl)-2-pyrrolidylmethylenebenzimidazole hydrochloride, 1-[(4-Chlorophenyl)methyl]-2-(1-pyrrolidinylmethyl)-1H-benzimidazole hydrochloride, CID 2782 hydrochloride, NSC 46261 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20ClN3 · HCl
CAS Number:
Molecular Weight:
362.30
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

ClC(C=C1)=CC=C1CN2C(CN3CCCC3)=NC4=CC=CC=C42.[H]Cl

InChI

1S/C19H20ClN3.ClH/c20-16-9-7-15(8-10-16)13-23-18-6-2-1-5-17(18)21-19(23)14-22-11-3-4-12-22;/h1-2,5-10H,3-4,11-14H2;1H

InChI key

DNFMJYXRIMLMBZ-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Clemizole is a potent and preferring inhibitor of TRPC5 that efficiently blocks heterologously expressed homomeric TRPC5 channels as well as heteromeric TRPC1:TRPC5 channels. Clemizole is a potent and selective H1 histamine receptor antagonist. Clemizole inhibited binding of HCV RNA by NS4B inhibit HCV RNA replication in cell culture.
H1 histamine receptor antagonist.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Oishi et al.
Naunyn-Schmiedeberg's archives of pharmacology, 349(2), 140-144 (1994-02-01)
To compare in vivo effects of eleven compounds of different classes of histamine H1-receptor antagonists (alcoholamines: diphenhydramine, carbinoxamine, and clemastine; ethylenediamines: mepyramine, tripelennamine, and clemizole; alkylamines: triprolidine and chlorpheniramine; piperazines: meclizine and homochlorcyclizine; phenothiazines: promethazine) on neuronal uptake of dopamine
Andreia Teixeira-Castro et al.
Brain : a journal of neurology, 138(Pt 11), 3221-3237 (2015-09-17)
Polyglutamine diseases are a class of dominantly inherited neurodegenerative disorders for which there is no effective treatment. Here we provide evidence that activation of serotonergic signalling is beneficial in animal models of Machado-Joseph disease. We identified citalopram, a selective serotonin
Jing Yuan et al.
Nature chemical biology, 5(10), 765-771 (2009-09-08)
Studies of gene function and molecular mechanisms in Plasmodium falciparum are hampered by difficulties in characterizing and measuring phenotypic differences between individual parasites. We screened seven parasite lines for differences in responses to 1,279 bioactive chemicals. Hundreds of compounds were
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service