Skip to Content
Merck
All Photos(1)

Documents

C2854

Sigma-Aldrich

Caspase 2 human

≥95% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution, >1000 units/mg protein

Synonym(s):

Ich-1, Nedd2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

buffered aqueous solution

specific activity

>1000 units/mg protein

mol wt

inactive proenzyme 56 kDa (active form consists of 17 kDa and 10 kDa subunits)
≥30 kDa

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CASP2(835)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Caspase 2 plays a major role in both positive and negative regulation of programmed cell death. It exists as two spliced variants, Ich-1L (long) and Ich-1S (short). Functionally, the two forms differ in that overexpression of the long form will induce apoptosis, while overexpression of the short form suppresses cell death.

Unit Definition

One unit will hydrolyze one nmol of N-Acetyl-Val-Asp-Val-Ala-Asp-AFC per min at pH 7.5 at 25 °C.

Physical form

Solution in 25 mM HEPES, pH 7.5, 0.15 M NaCl, 5 mM dithiothreitol, 10% sucrose, with 0.5 mg bovine serum albumin

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Wang et al.
Cell, 78(5), 739-750 (1994-09-09)
We report here the isolation and characterization of Ich-1, a gene related to the C. elegans cell death gene ced-3 and the mammalian homolog of ced-3, interleukin-1 beta-converting enzyme (ICE). Alternative splicing results in two distinct Ich-1 mRNA species. One

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service