C-21060
Airway Epithelial Cell Growth Medium
Ready-to-use kit including Basal Medium and SupplementMix, 500 ml
Synonym(s):
Primary Cell Culture Media
About This Item
Recommended Products
form
aqueous solution
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
General description
Application
Components
Quality
Preparation Note
Storage and Stability
Subculture Routine
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Optimize primary human airway epithelial cell culture with serum-free media and handling protocols for large air passage cells.
Related Content
Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service