Key Documents
BAF134690003
Bel-Art® Write-on Tape Dispenser
Separator Discs
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
packaging
pack of 3
manufacturer/tradename
Bel-Art F13469-0003
General description
Prevents adjacent rolls of labeling tape from binding with each other. Use when loading dispenser with more than 4 rolls of labeling tape.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)