Skip to Content
Merck
All Photos(1)

Key Documents

B9686

Sigma-Aldrich

Monoclonal Anti-Brain-derived Neurotrophic Factor antibody produced in mouse

clone 37129, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-BDNF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

37129, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: 2 μg/mL

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... BDNF(627)

Looking for similar products? Visit Product Comparison Guide

General description

Brain derived neurotrophic factor (BDNF) belongs to a family of secreted proteins called neurotrophins that includes, nerve growth factor (NGF), neurotrophin-3 (NT3) and NT4/5. BDNF is expressed in the developing and the mature brain. The influence of BDNF is mediated by its binding to TrkB receptor and sometimes the receptor p75NTR. The downstream pathways promoted by BDNF include PI3K, MAPK and JAK/STAT. The most important role of BDNF is the regulation of synaptic transmission and plasticity, protein synthesis and phosphorylation of local proteins. The secretion of BDNF is highly regulated at both transcriptional and translational levels since it is involved in the proper function of neuronal circuit and development of cognitive functions. The deficits in BDNF functions are implicated in a number of neurodegenerative disorders such as Huntington′s disease, schizophrenia and dementia
Monoclonal Anti-Brain-derived Neurotrophic Factor detects human BDNF.

Immunogen

purified, Sf21-derived recombinant human brain-derived neurotrophic factor.

Application

Monoclonal Anti-Brain-derived Neurotrophic Factor may be used to detect human BDNF by ELISA using a working antibody concentration of 2 μg/ml.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brain-derived neurotrophic factor and the development of structural neuronal connectivity
Cohen-Cory S et al
Developmental neurobiology, 70, 271-278 (2012)
Jordi Alberch et al.
Progress in brain research, 146, 195-229 (2004-01-01)
Huntington's disease is a neurodegenerative disorder characterized by the selective loss of striatal neurons and, to a lesser extent, cortical neurons. The neurodegenerative process is caused by the mutation of huntingtin gene. Recent studies have established a link between mutant
Mark P Mattson et al.
Trends in neurosciences, 27(10), 589-594 (2004-09-18)
Brain-derived neurotrophic factor (BDNF) and serotonin (5-hydroxytryptamine, 5-HT) are known to regulate synaptic plasticity, neurogenesis and neuronal survival in the adult brain. These two signals co-regulate one another such that 5-HT stimulates the expression of BDNF, and BDNF enhances the
Hai-Yang Zhang et al.
Asian journal of andrology, 13(2), 231-235 (2010-12-21)
Retroperitoneal operations, such as radical prostatectomy, often damage the cavernous nerve, resulting in a high incidence of erectile dysfunction. Although improved nerve-sparing techniques have reduced the incidence of nerve injury, and the administration of phosphodiesterase type 5 inhibitors has revolutionized
Bai Lu et al.
Novartis Foundation symposium, 289, 119-129 (2008-05-24)
BDNF is a key regulator of synaptic plasticity and hence is thought to be uniquely important for various cognitive functions. While correlations of schizophrenia with polymorphisms in the BDNF gene and changes in BDNF mRNA levels have been reported, specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service