Skip to Content
Merck
All Photos(1)

Key Documents

B7885

Sigma-Aldrich

[Tyr8]-Bradykinin

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H73N15O12
CAS Number:
Molecular Weight:
1076.21
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

NC(CCCNC(N)=N)C(=O)N1CCCC1C(=O)N2CCCC2C(=O)NCC(=O)NC(Cc3ccccc3)C(=O)NC(CO)C(=O)N4CCCC4C(=O)NC(Cc5ccc(O)cc5)C(=O)NC(CCCNC(N)=N)C(O)=O

Amino Acid Sequence

Arg-Pro-Pro-Gly-Phe-Ser-Pro-Tyr-Arg

Application

Useful for determination of human urinary kinin levels

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of human urinary kinin levels by radioimmunoassay using a tyrosine analogue of bradykinin.
M L Mashford et al.
Biochemical pharmacology, 20(5), 969-973 (1971-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service