Skip to Content
Merck
All Photos(1)

Key Documents

B0643

Sigma-Aldrich

Boc-Leu-OH

Synonym(s):

Boc-L-leucine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
231.29
Beilstein:
1711814
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C11H21NO4/c1-7(2)6-8(9(13)14)12-10(15)16-11(3,4)5/h7-8H,6H2,1-5H3,(H,12,15)(H,13,14)/t8-/m0/s1

InChI key

MDXGYYOJGPFFJL-QMMMGPOBSA-N

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huang-Han Chen et al.
Analytica chimica acta, 865, 53-59 (2015-03-04)
Polydimethylsiloxane (PDMS) is widely used for microfabrication and bioanalysis; however, its surface functionalization is limited due to the lack of active functional groups and incompatibility with many solvents. We presented a novel approach for in situ fabrication of cleavable peptide
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service