Skip to Content
Merck
All Photos(1)

Key Documents

AXYVT38431ULR

Pipette tips in 384-tip format, robotic

size 30 μL, pkg of 5x3840tips/cs, racked

Synonym(s):

Pipette tips for Velocity

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
37.520,00 kr.

37.520,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
37.520,00 kr.

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
30 μL
sterility:
non-sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 5x3840tips/cs
manufacturer/tradename:
Axygen Inc VT-384-31UL-R
Corning

37.520,00 kr.


Please contact Customer Service for Availability

material

clear

sterility

non-sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no

manufacturer/tradename

Axygen Inc VT-384-31UL-R
Corning

packaging

pkg of 5x3840tips/cs

size

30 μL

volume

30 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® automation pipette tips in 384-tip format are designed for Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST, and 16ST head.
  • Manufactured to stringent specifications including automated assembly to minimize contamination risks and ensure consistent performance and quality.
  • The clear polypropylene high-quality tips are ideal for sensitive biological applications.
  • RNase-/DNase-free and nonpyrogenic.

Other Notes

Browse all pipette tips

Legal Information

Agilent is a registered trademark of Agilent Technologies, Inc.
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Velocity11 VPrep is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service