AXYUC500
Pressure sensitive sealing film, ultra clear, polyester, 70 μm
pkg of 5x100sheets/cs
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
Select a Size
15.760,00 kr.
Select a Size
About This Item
15.760,00 kr.
Recommended Products
material
polyolefin
sterility
non-sterile
feature
Ideal for protaincrystallography and other applications requiring ultimate optical clarity
manufacturer/tradename
Axygen Inc UC-500
Corning
packaging
pkg of 5x100sheets/cs
Related Categories
Features and Benefits
- Easy to use
- Cost-effective
- Suitable for 96 and 384 well PCR microplates
- Film clarity facilitates optical analysis during real-time PCR
- Contains pressure-sensitive adhesive which avoids tacky residues that can interfere with optical analysis
- Withstands temperature ranging from -70°C to 100°C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service