AXYTR222CL
MAXYMum Recovery® pipette reference tips
universal fit, size 200 μL, pkg of 20x1000tips/cs
Synonym(s):
pipet tips
Select a Size
5.200,00 kr.
Select a Size
About This Item
RNase free
barrier: no
endotoxin free
filter: no
graduation
Corning
5.200,00 kr.
Recommended Products
material
clear
sterility
non-sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter: no
graduation
manufacturer/tradename
Axygen Inc TR-222-C-L
Corning
packaging
pkg of 20x1000tips/cs
size
200 μL
volume
200 μL
Looking for similar products? Visit Product Comparison Guide
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service