AXYSCT050RS
Screw cap tubes and caps with O-rings, conical bottom
capacity 0.5 mL, red, sterile
Synonym(s):
centrifuge tubes
Select a Size
7.840,00 kr.
Select a Size
About This Item
7.840,00 kr.
Recommended Products
material
red
screw top cap
sterility
sterile
feature
RCF 20,000 × g
packaging
pack of 500 tubes
case of 8 packs
manufacturer/tradename
Axygen Inc
Corning
capacity
0.5 mL
Choose from one of the most recent versions:
Certificates of Analysis (COA)