AXYSCOC
Screw caps with O-rings
clear, pkg of 8x500caps/cs
Synonym(s):
Caps with O-Rings, Screw Caps
Select a Size
7.120,00 kr.
Select a Size
About This Item
7.120,00 kr.
manufacturer/tradename
Axygen Inc SCO-C
Corning
packaging
pkg of 8x500caps/cs
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service