AXYPCR384RGDC
Corning® Axygen® 384 Well RigiPlate® PCR Microplate
Full Skirt, size 384 wells, clear, conical bottom clear polypropylene wells, pkg of 5x20plates/cs
Synonym(s):
PCR Rigi plate microplate
Select a Size
8.640,00 kr.
Select a Size
About This Item
8.640,00 kr.
material
conical bottom clear polypropylene wells
sterility
non-sterile
feature
skirt
manufacturer/tradename
Axygen Inc PCR-384-RGD-C
Corning
packaging
pkg of 5x20plates/cs
maximum volume
49.5 μL
size
384 wells
working volume
49.5 μL
color
clear
suitability
suitable for PCR
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service