Skip to Content
Merck
All Photos(1)

Key Documents

AXYMCT500C

Corning® Axygen® MaxyClear Snaplock Microcentrifuge Tube

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
3.420,00 kr.

3.420,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
3.420,00 kr.

About This Item

UNSPSC Code:
41121703
NACRES:
NB.15

3.420,00 kr.


Please contact Customer Service for Availability

material

cap (snap cap)
conical bottom tube
polypropylene

sterility

non-sterile

feature

RNase and DNase free
endotoxin free

packaging

case of 1250 ea
pack of 250 ea

manufacturer/tradename

Corning MCT-500-C

parameter

14000 × g max. RCF

color

clear

capacity

5 mL

fitting

cap (push fit)

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Made up of homopolymer polypropylene with graduations at 0.5 mL intervals, frosted cap surfaces for labeling, and frosted panels on the side for writing
  • Clear/amber microtubes that can withstand relative centrifugal force (RCF) up to 14,000
  • Ensures efficient seal due to snap lock cap
  • Free from RNase and DNase for complete sample integrity
  • Nonpyrogenic in nature

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service