Skip to Content
Merck
All Photos(1)

Documents

AV46056

Sigma-Aldrich

Anti-EIF2S3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-EIF2, Anti-EIF2G, Anti-EIF2gamma, Anti-Eukaryotic translation initiation factor 2, subunit 3 γ, 52 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

human, dog, rat, bovine, rabbit, guinea pig, mouse, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EIF2S3(1968)

Immunogen

Synthetic peptide directed towards the N terminal region of human EIF2S3

Application

Anti-EIF2S3 antibody produced in rabbit is suitable for western blotting at a concentration of 5μg/ml.

Biochem/physiol Actions

EIF2S3 gene encodes a eukaryotic translation initiation factor 2, subunit 3 gamma protein of 52kDa. It is the core subunit of the heterotrimeric GTP-binding protein that facilitates the recruitment of methionyl-tRNA to the 40S ribosomal subunit.

Sequence

Synthetic peptide located within the following region: LDDPSCPRPECYRSCGSSTPDEFPTDIPGTKGNFKLVRHVSFVDCPGHDI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guntram Borck et al.
Molecular cell, 48(4), 641-646 (2012-10-16)
Together with GTP and initiator methionyl-tRNA, translation initiation factor eIF2 forms a ternary complex that binds the 40S ribosome and then scans an mRNA to select the AUG start codon for protein synthesis. Here, we show that a human X-chromosomal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service