Skip to Content
Merck
All Photos(2)

Key Documents

AV44054

Sigma-Aldrich

Anti-SLC25A28 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp547C109, Anti-MFRN2, Anti-MRS3/4, Anti-MRS4L, Anti-NPD016, Anti-Solute carrier family 25, member 28

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
3.490,00 kr.

3.490,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
3.490,00 kr.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

3.490,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the C terminal region of human SLC25A28

Application

Anti-SLC25A28 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

SLC25A28 also known as mitoferrin 2 (MFRN2) belongs to mitochondrial solute carrier family.It transports mitochondrial iron into the developing erythrocytes. High concentrations of iron are highly toxic; the regulation of iron transport in the vertebrate cells by MFRN1 and 2 is therefore critical.

Sequence

Synthetic peptide located within the following region: NTQESLALNSHITGHITGMASAFRTVYQVGGVTAYFRGVQARVIYQIPST

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prasad N Paradkar et al.
Molecular and cellular biology, 29(4), 1007-1016 (2008-12-17)
Mitoferrin 1 and mitoferrin 2 are homologous members of the mitochondrial solute carrier family. Mitoferrin 1 is required for mitochondrial iron delivery in developing erythrocytes. Here we show that mitoferrin 1 and mitoferrin 2 contribute to mitochondrial iron delivery in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service