Skip to Content
Merck
All Photos(1)

Key Documents

AV43761

Sigma-Aldrich

Anti-SLC25A25 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-KIAA1896, Anti-MCSC, Anti-MGC105138, Anti-PCSCL, Anti-SCAMC-2, Anti-Solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 25

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
4.100,00 kr.

4.100,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
4.100,00 kr.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

4.100,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

horse, human, bovine, dog, rabbit, guinea pig, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC25A25

Biochem/physiol Actions

SLC25A25 is a member of family of calcium-binding mitochondrial carriers located in the inner membranes of mitochondria. It is ATP-Mg/Pi carrier and regulates the movement of adenine nucleotides in mitochondria. SLC25A25 maintains the homeostasis of ATP and Ca+2 cycling in the skeletal muscle.

Sequence

Synthetic peptide located within the following region: AVGLRRLGLHRTEGELQKIVQAGDKDLDGQLDFEEFVHYLQDHEKKLRLV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rea P Anunciado-Koza et al.
The Journal of biological chemistry, 286(13), 11659-11671 (2011-02-08)
An ATP-Mg(2+/)P(i) inner mitochondrial membrane solute transporter (SLC25A25), which is induced during adaptation to cold stress in the skeletal muscle of mice with defective UCP1/brown adipose tissue thermogenesis, has been evaluated for its role in metabolic efficiency. SLC25A25 is thought
Araceli del Arco et al.
The Journal of biological chemistry, 279(23), 24701-24713 (2004-04-01)
Aralar1 and citrin were identified as calcium binding aspartate/glutamate carriers (AGC) in mitochondria. The presence of calcium binding motifs facing the extramitochondrial space allows the regulation of the transport activity of these carriers by cytosolic calcium and provides a new

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service