All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H25N5O5
CAS Number:
Molecular Weight:
427.45
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
Recommended Products
Assay
≥98% (HPLC)
form
powder
solubility
0.1% trifluoroacetic acid in acetonitrile: water (3:1): 1 mg/mL, clear, colorless
storage temp.
−20°C
SMILES string
CC(N)C(=O)NC(C)C(=O)NC(Cc1ccccc1)C(=O)Nc2ccc(cc2)N(=O)=O
InChI
1S/C21H25N5O5/c1-13(22)19(27)23-14(2)20(28)25-18(12-15-6-4-3-5-7-15)21(29)24-16-8-10-17(11-9-16)26(30)31/h3-11,13-14,18H,12,22H2,1-2H3,(H,23,27)(H,24,29)(H,25,28)
InChI key
RMYMOVDNWWDGFU-UHFFFAOYSA-N
Application
Substrate for chymotrypsin and tripeptidyl peptidase I.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Robert Dalton et al.
International journal of molecular sciences, 22(3) (2021-01-31)
Dendritic cells (DCs) are professional antigen presenting cells with a great capacity for cross-presentation of exogenous antigens from which robust anti-tumor immune responses ensue. However, this function is not always available and requires DCs to first be primed to induce
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service