Skip to Content
Merck
All Photos(1)

Key Documents

A8728

Sigma-Aldrich

Anti-phospho-β-Arrestin (pSer412) antibody produced in rabbit

affinity isolated antibody, aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
6.150,00 kr.

6.150,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
6.150,00 kr.

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

6.150,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

aqueous solution

species reactivity

rat, mouse

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

β-arrestin-1 is a scaffold protein that is highly expressed in the central nervous system. This adaptor protein regulates mitogenic signaling, and also mediates the desentization and internalization of G protein coupled receptors (GPCRs).
Rabbit Anti-phospho-β-Arrestin (pSer412) antibody detects mouse and rat β-arrestin-1. The antibody is also expected to react with human and cow antigens due to the high degree of homology (77%).

Immunogen

synthetic phosphopeptide derived from the region of rat β-arrestin-1 that contains serine 412.

Application

Rabbit Anti-phospho-beta-Arrestin (pSer412) antibody can be used for western blot assays.

Physical form

Solution in Dulbecco′s PBS, pH 7.3 with 1 mg/mL BSA and 0.05% Sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service