Skip to Content
Merck
All Photos(1)

Documents

A6762

Sigma-Aldrich

Aerobacter aerogenes cells

Type III, Lyophilized cells grown on a medium containing inositol

Synonym(s):

Enterobacter aerogenes

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204

sterility

(not processed or packaged aseptically)

type

Type III

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Crude source of inositol dehydrogenase.
Not intended for use as a starter culture.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sigifredo Pedraza-Sánchez et al.
International immunopharmacology, 6(4), 635-646 (2006-03-01)
The compound RU41740 from Klebsiella pneumoniae, when used as an immunostimulant, improves responses to bacterial and yeast infections in murine models and in human trials. The aim of this study was to determine in vitro, the capacity of RU41740 to
J J Finlay-Jones et al.
Journal of medical microbiology, 38(6), 454-458 (1993-06-01)
The prophylactic and therapeutic efficacies of the immunomodulating agent RU 41.740 (a glycoprotein extract from Klebsiella pneumoniae) were studied in a murine model of intra-abdominal abscess formation with Bacteroides fragilis, Escherichia coli, and bran as an abscess-potentiating agent. Parenteral injection
B de Wazières et al.
Immunopharmacology, 39(1), 51-59 (1998-07-17)
The aim of this study was to analyze the effects of stress on natural immunity in old mice, and the potential of an immunomodulating drug to correct stress-induced immune abnormalities. We analyzed both the alveolar (ALM) and peritoneal macrophage (PerM)
M Roch-Arveiller et al.
Inflammation, 15(6), 437-445 (1991-12-01)
An impairment of polymorphonuclear leukocyte (PMN) functions has been described following burn trauma. It was thus of interest to investigate the effect of RU 41740, an agent known to stimulate these cells, on rat PMN functions after burn injury. In
H Blomgen et al.
Anticancer research, 11(5), 1849-1853 (1991-09-01)
Biostim, which is a glucoprotein extract of Klebsiella pneumoniae, is known to trigger human monocytes to increased secretion of metabolites, which inhibit lymphocyte mitogenesis and which augment NK-activity of lymphocytes. Some aspects of this monocyte activation have been examined in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service