All Photos(1)
About This Item
Linear Formula:
(CD3CO)2O
CAS Number:
Molecular Weight:
108.13
Beilstein:
1910689
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
refractive index
n20/D 1.3875 (lit.)
bp
138-140 °C (lit.)
mp
-73 °C (lit.)
mass shift
M+6
SMILES string
[2H]C([2H])([2H])C(=O)OC(=O)C([2H])([2H])[2H]
InChI
1S/C4H6O3/c1-3(5)7-4(2)6/h1-2H3/i1D3,2D3
InChI key
WFDIJRYMOXRFFG-WFGJKAKNSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
120.2 °F - closed cup
Flash Point(C)
49 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service