All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H15N5O4
CAS Number:
Molecular Weight:
293.28
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Assay
≥98%
storage temp.
−20°C
SMILES string
CC(=O)OC1CC(OC1CO)n2cnc3c(N)ncnc23
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service