Skip to Content
Merck
All Photos(1)

Key Documents

A3435

Sigma-Aldrich

D-Alanine–Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

form

saline suspension

extent of labeling

2-8 μmol per mL

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Application

D-Alanine Agarose can be used in proteomics and protein chromatography. It has been utilized in experiments with antibiotics of the vancomycin group to determine the role of micellular aggregates in the high binding affinity of teicoplanin to D-alanyl-D-alanine-agarose.

Physical form

Suspension in 2.0 M NaCl containing preservative

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Corti et al.
Clinical chemistry, 31(10), 1606-1610 (1985-10-01)
A solid-phase enzyme-receptor assay (SPERA) has been developed for glycopeptide antibiotics of the vancomycin class such as teicoplanin, vancomycin, ristocetin, avoparcin, actaplanin, A-47934, A-41030, and A-35512-B. The assay exploits the mechanism of most action of these antibiotics, which is based
A Corti et al.
Journal of applied biochemistry, 7(2), 133-137 (1985-04-01)
Teicoplanin, as well as the other antibiotics of the vancomycin group, was shown to bind specifically to D-alanyl-D-alanine-agarose (D-Ala-D-Ala-AGA) (A. Corti and G. Cassani, Appl. Biochem. Biotechnol. 11, 101-110 (1985)). This finding is extended, showing that the binding is as
Catherine F Silverio et al.
Electrophoresis, 24(5), 808-815 (2003-03-11)
Binding constants between the glycopeptides teicoplanin (Teic) and ristocetin (Rist) and their derivatives to D-Ala-D-Ala terminus peptides were determined by on-column receptor synthesis coupled to partial-filling affinity capillary electrophoresis (PFACE) or affinity capillary electrophoresis (ACE). In these techniques, the column
Makoto Umeda et al.
Biochemical and biophysical research communications, 466(4), 717-722 (2015-09-09)
Arginine, a semi-essential amino acid, is known as one of the most strongest insulin secretagogues in a glucose-dependent manner, but major mechanism is unknown. Arginine induced insulin secretion in mice as well as β cell line, NIT-1, in which more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service