Skip to Content
Merck
All Photos(1)

Key Documents

A2727

Sigma-Aldrich

N-Acetyl-Adhesin Amide Fragment 1025-1044

≥95% (HPLC)

Synonym(s):

p1025

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C97H160N26O32
Molecular Weight:
2202.46
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.26

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

−20°C

Amino Acid Sequence

Ac-Gln-Leu-Lys-Thr-Ala-Asp-Leu-Pro-Ala-Gly-Arg-Asp-Glu-Thr-Thr-Ser-Phe-Val-Leu-Val-NH2

Biochem/physiol Actions

Antimicrobial peptide that has been shown to specifically inhibit binding of a cell surface adhesion of Streptococcus mutans, the main etiological agent of dental caries, to salivary receptors in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C G Kelly et al.
Nature biotechnology, 17(1), 42-47 (1999-01-27)
The earliest step in microbial infection is adherence by specific microbial adhesins to the mucosa of the oro-intestinal, nasorespiratory, or genitourinary tract. We inhibited binding of a cell surface adhesin of Streptococcus mutans to salivary receptors in vitro, as measured

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service