Skip to Content
Merck
All Photos(1)

Documents

A1413

Sigma-Aldrich

Anisodine hydrobromide

Synonym(s):

α-Hydroxyscopolamin hydrobromide, Daturamine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO5 · HBr
CAS Number:
Molecular Weight:
400.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

SMILES string

Br.CN1[C@H]2CC(C[C@@H]1[C@@H]3O[C@@H]23)OC(=O)C(O)(CO)c4ccccc4

InChI

1S/C17H21NO5.BrH/c1-18-12-7-11(8-13(18)15-14(12)23-15)22-16(20)17(21,9-19)10-5-3-2-4-6-10;/h2-6,11-15,19,21H,7-9H2,1H3;1H/t11?,12-,13+,14-,15+,17-;/m1./s1

InChI key

GJPDCORRBGIJOP-KALLRANASA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Run-sheng Wang et al.
[Zhonghua yan ke za zhi] Chinese journal of ophthalmology, 46(12), 1079-1083 (2011-01-08)
To investigate changes of plasma endothelin-1 (ET-1) concentration in photodynamic induced rat anterior ischemic optic neuropathy model (rAION) and evaluate the effects of compound anisodine hydrobromide (CA). Eighty-five Sprague-Dawley (SD) male rats were randomly divided into a blank control group
W Xu et al.
Zhongguo yao li xue bao = Acta pharmacologica Sinica, 17(2), 161-163 (1996-03-01)
To study the protective effect of anisodine (Ani) on acute forebrain ischemia-reperfusion injury in rats. Both vertebral arteries were occluded by electrocautery. Severe, but transient bilateral cerebral ischemia was produced by clamping both common carotid arteries in rats. Atomic absorption
D C Thompson et al.
Mutation research, 279(2), 83-89 (1992-05-16)
In previous studies on the mutagenicity of anisidine isomers, the ortho isomer was considered to be mutagenic towards standard Ames tester strains, while the para isomer gave equivocal results. In the present study we show that both para- and ortho-anisidine
Huaixia Chen et al.
Journal of pharmaceutical and biomedical analysis, 44(3), 773-778 (2007-04-17)
Liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESIMSn) was employed to investigate the in vivo and in vitro metabolism of anisodine. Feces, urine and plasma samples were collected after ingestion of 20 mg anisodine to healthy rats. Feces and urine samples
Shao-jie Shan et al.
Phytotherapy research : PTR, 16(5), 449-454 (2002-08-31)
The effects of Yi Zhi Fang extract (YZF) on learning and memory performances were investigated in mice using passive avoidance tasks. Oral administration of YZF improved learning and memory disorders induced by chemicals in both normal and senile mice. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service